Learn More
Invitrogen™ Recombinant Proteinase K Solution (20 mg/mL)
Recombinant Proteinase K is a nonspecific serine protease that is useful for general digestion of proteins.
Brand: Invitrogen™ AM2546
Description
Recombinant Proteinase K is a nonspecific serine protease that is useful for general digestion of proteins.
Proteinase K remains active:
• Over a wide pH range (optimal activity between 6.5 and 9.5)
• Under denaturing conditions (e.g., in the presence of SDS or urea)
• In the presence of metal chelating agents (e.g., EDTA)
• At comparatively high temperatures (optimum digestion temperature is 65°C)
Applications
Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.
Performance and quality testing
Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.
Unit definition
One mAnson unit is described as that amount of enzyme that liberates 1 μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.
Order Info
Shipping Condition: Dry Ice
Specifications
20 mg/mL | |
Store at -20°C. Shipped at ambient temperature. |
|
1.25 mL | |
Ambion™ |
T. album | |
Dry Ice | |
Proteinase K | |
Liquid |
Safety and Handling
- PROTEINASE K (20MG/ML) 1.25ML
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Proteinase, tritirachiumalbum serine
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.