missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RiboPure™ RNA Purification Kit

The Ambion™ RiboPure™ Kit is designed for rapid purification of high-quality RNA from tissue samples or cultured cells. The kit combines TRI Reagent™ with glass fiber filter purification to yield exceptionally pure RNA, free of residual proteins and lipids.

790.00 € - 924.00 €

Specifications

Isolation Technology Spin Column (Glass Fiber Filter)
Purification Time 30 min. (After sample homogenization)
Elution Volume 100 μL
Sample Type Cells, Tissue
Final Product Type Total RNA
View More Specs

Products 2
Product Code Brand Product Type Includes Price Quantity & Availability  
Product Code Brand Product Type Includes Price Quantity & Availability  
10321325
View Documents Promotion Details
Invitrogen™
AM1924M
Kit with manual TRI reagent, filter cartridges, 2 mL collection tubes, wash solution concentrate, elution buffer, and manual This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10107824
View Documents Promotion Details
Invitrogen™
AM1924
790.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Ambion™ RiboPure™ Kit is designed for rapid purification of high-quality RNA from tissue samples or cultured cells. The kit combines TRI Reagent™ with glass fiber filter purification to yield exceptionally pure RNA, free of residual proteins and lipids. The kit includes sufficient reagents for 50 purifications from 1–100 mg of tissue or 0.1–20 x 106 cultured cells each.

• Yields the highest purity RNA attainable
• Ideal for the most stringent downstream applications
• Handles problematic samples with ease
• Fast (less than 30 minutes) and easy procedure

RNA isolated using the RiboPure™ Kit is suitable for use in all common downstream applications, including cDNA synthesis, real-time and end-point RT-PCR, microarray analysis, northern blots, and RNase protection assays. The RiboPure™ protocol is especially suited for processing samples that are difficult to disrupt, or samples that have large amounts of ribonuclease or other contaminants. Samples are first homogenized in TRI Reagent™, a monophasic solution containing phenol and guanidine thiocyanate, to rapidly lyse cells and inactivate nucleases. Addition of bromochloropropane (BCP) results in separation of the homogenate into aqueous and organic phases. RNA partitions to the aqueous phase, while DNA and protein remain in the interphase and organic phase. The aqueous phase is removed and passed through a glass fiber filter that binds the RNA. The filter is washed, and the RNA is eluted. The procedure is compatible with tissues that have been stored in Ambion's RNAlater™. Total RNA yield is typically 100–500 μg per 100 mg of tissue, depending on the type of tissue.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Spin Column (Glass Fiber Filter)
100 μL
Total RNA
RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Cells: 0.1–20 x 106
Tissue: 5–100 mg
30 min. (After sample homogenization)
Cells, Tissue
Not High-throughput Compatible (Manual)
Room Temperature
Up to 500 μg
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.