missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNase Cocktail™ Enzyme Mix

Endonuclease-free, exonuclease-free, protease-free

Brand:  Invitrogen™ AM2286

174.80 EUR valid until 2025-12-31
Use promo code "24274" to get your promotional price.


 View more versions of this product

Product Code. 10036284

  • 192.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

RNase Cocktail™ is mixture of two highly purified ribonucleases, RNase A (500U/mL) and RNase T1 (20,000U/mL) and is free of DNase and nicking activities. Digestion of RNA with RNase A alone leaves fragments of RNA which are large enough to be visible on agarose gels and precipitate in ethanol. RNase A cuts after C and U residues, and RNase T1 cuts after G residues. Consequently, mixture of both enzymes results in reduction in RNA fragment size over use of either alone.

  • Use RNase Cocktail for all situations, desirable to degrade RNA, i.e. plasmid minipreps and ribonuclease protection assays
  • RNase Cocktail is supplied in 50% glycerol for maximum convenience
  • Unit concentration: RNase A at 500U/mL and RNase T1 at 20,000U/mL
  • Quality Control: RNase Cocktail Enzyme Mix is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and protease activity; functionality is determined in ribonuclease protection assay
  • Unit Definitions: RNase A: One unit of RNase A is amount required to give increase in absorption at 286nm of 0.0146 absorbance units per minute in a 1mL volume
  • Unit assay conditions: 100mM Tris-acetate (pH 6.5), 1mM EDTA and 1mM cyclic 2', 3'-CMP
  • RNase T1: 100 Units of RNase T1 is amount of enzyme that yields an increase in absorption at 260nm of 0.01428 units per min. at room temperature using 60μg/mL yeast total RNA as a substrate

DNA & RNA Purification & Analysis, DNA Extraction, Maxiprep, Midiprep, Miniprep, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting, Plasmid DNA Purification

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

RNase A = 500 U/mL, RNase T1 = 20,000 U/mL
Ambion™ RNase Cocktail™ is an enzyme mix which contains two highly purified ribonucleases, RNase A and RNase T1. Unit concentration is: RNase A at 500 U⁄ml and RNase T1 at 20,000 U⁄ml.
Store at -20°C. Do not store in a frost-free freezer.
RNase
RNAse Cocktail Enzyme Mix
Endonuclease-Free, Protease-Free, Exonuclease-Free
Dry Ice
1 mL
Ambion™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ RNase Cocktail™ Enzyme Mix > 1mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.