Learn More
Invitrogen™ RNase-free Tips, 1,000 μL size
Provided in ten (10) racks containing 100 tips each. Invitrogen™ Ambion™ RNase-free Tips, 1000μL size are subjected to rigorous testing for RNase contamination and are certified to be RNase- and DNase-free.
Brand: Invitrogen™ AM12660
159.60 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Invitrogen™ Tips are compatible with Eppendorf™, Finnpipette, Oxford Benchmate™, Pipetman™, Proline™, Rainin, Socorex, and other brands of pipettors.
Specifications
1000 μL | |
Sterile | |
Store at room temperature. | |
Universal Fit |
Non-filtered | |
RNase-free Tips | |
Room Temperature | |
Ambion |