Learn More
Invitrogen™ Ambion™ RNase I, cloned, 100 U/μL
Efficiently cleaves after all four bases of single-stranded RNA, in contrast to RNase A, which only cleaves after C and U residues
275.00 € - 492.00 €
Specifications
Concentration | 100 U/μL |
---|---|
Content And Storage | -20°C |
Shipping Condition | Dry Ice |
Enzyme | RNase |
Product Type | RNase I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10545195
|
Invitrogen™
AM2294 |
10,000 U |
275.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10330065
|
Invitrogen™
AM2295 |
25,000 U |
492.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNase I degrades all RNA dinucleotide bonds leaving a 5' hydroxyl and 2', 3' cyclic monophosphate. RNase I will degrade any RNA to mixture of mononucleotides, dinucleotides, and trinucleotides and does not degrade DNA, although it will bind to DNA. It has marked preference for single-stranded RNA over double-stranded RNA, which allows it to work well in ribonuclease protection assays.
- Supplied in one tube of 10,000U (100U/μL)
- Endonuclease-free, exonuclease-free, protease-free
- Rigorously tested for nonspecific contaminating endonuclease, exonuclease, and protease activity
- Unit Definition: One unit is amount of enzyme required to produce 1μg of acid-soluble material from mouse liver RNA in 30 min. at 37°C
DNA & RNA Purification & Analysis, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting
Order Info
Shipping Conditions: Dry ice
Specifications
100 U/μL | |
Dry Ice | |
RNase I |
-20°C | |
RNase | |
Ambion™ |
For Research Use Only. Not for use in diagnostic procedures.