missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ RNase Inhibitor, cloned, 40 U/μL

Binds neutral pancreatic RNase A type enzymes

Brand:  Invitrogen™ AM2682

195.96 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


 View more versions of this product

Product Code. 10177094

  • 276.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ambion RNase Inhibitor, a recombinant human protein produced in E. coli, is a potent inhibitor of neutral pancreatic RNase A type enzymes. The mode of inhibition is noncompetitive; the inhibitor tightly binds RNases in a 1:1 ratio. The enzyme has been shown to inactivate RNases present in many tissues and cell types. Addition of the ribonuclease inhibitor (RI) has been shown to be useful whenever the integrity of RNA must be maintained such as in the preparation of cDNA by reverse transcription, in vitro RNA transcription, and in in vitro protein synthesis. RI does not inhibit RNase I, T1, T2, H, U1, U2, or CL3. RI requires a minimum of 1mM DTT to maintain activity, and has an activity range between pH 5.0 to 8.0 with maximal activity between pH 7.0 and 8.0. Since the mode of inhibition is the formation of a 1:1 complex with RNases, care must be taken to avoid denaturation or oxidation of the ribonuclease inhibitor (e.g., by addition of SDS, urea, etc.). RI should be added to transcription, translation, and cDNA synthesis reactions to give a final concentration of 1U/μL. RNase Inhibitor is rigorously tested for contaminating RNase, exonuclease, endonuclease, and protease activity.
Unit Definition: One unit is the amount of protein required to inhibit the activity of 5ng of RNase A by 50%. Unit assay conditions: 100mM Tris-acetate (pH 6.5), 1mM EDTA, 1mM cyclic 2 ft.,3 ft.-CMP, and RNase inhibitor. Addition of RNase A initiates the reaction. RNase Inhibitor activity is measured by the inhibition of hydrolysis of cyclic 2 ft., 3 ft.-CMP by RNase A.

Applications:

Gene Expression Analysis and Genotyping, In Vitro Transcription, PCR and Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mM
Ambion™
Dry Ice
Store at -20°C. Do not store in a frost–free freezer.
2500 U
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Ambion™ RNase Inhibitor, cloned, 40 U/μL > 2500 units

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.