missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNaseAlert™ Lab Test Kit v2

RNaseAlert™ Lab Test Kit v2 detects RNase activity in a convenient and sensitive fluorimetric assay that delivers results in real time.

Brand:  Invitrogen™ 4479768

Product Code. 15276535

  • 422.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

RNaseAlert™ Lab Test Kit v2 detects RNase activity in a convenient and sensitive fluorimetric assay that delivers results in real time. This kit is suitable for testing small numbers of sample and ensuring that solutions, tubes, tips, etc. in your lab are RNase-free. This new version contains a more sensitive substrate that is ∼60% more sensitive than the previous version. The kit contains sufficient reagents for 25 reactions.

  • Detects as little as approx. 0.3 pg of RNase A with a fluorometer
  • Simple, straight-forward 30-minute assay
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at -20°C:
• 25 tubes of RNaseAlert™ Substrate v2
• 250 μL 10X RNaseAlert Lab Test Buffer
• 500 μL RNase A

Store at room temperature:
• 50 mL RNaseZap™ Solution

Store at any temperature:
• 1.75 mL Nuclease-free Water

RNase Control Reagent
RNA Extraction
25 reactions
RNaseAlert™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2860142 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number4479768
Lot Number2803905 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4479768
Lot Number2803915 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4479768
Lot Number2828174 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4479768
Lot Number2820484 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4479768
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ RNaseAlert™ Lab Test Kit v2 > 25 reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.