Learn More
Invitrogen™ SDS, 20% Solution, RNase-free
Invitrogen molecular biology grade, SDS, 20% Solution is supplied in one bottle containing 250 mL.
Brand: Invitrogen™ AM9820
94.76 EUR valid until 2024-07-31
Use promo code "21860" to get your promotional price.
Additional Details : Weight : 0.10500kg
Description
Invitrogen molecular biology grade, SDS, 20% Solution is supplied in one bottle containing 250 mL. The buffer is certified RNase-free, economical, and ready-to-use. SDS is used as a component of both hybridization and wash buffers for northern and Southern blots and has many other uses around the lab.
Order Info
Shipping Condition: Room temperature
Specifications
Store at room temperature. | |
SDS | |
Molecular Biology Grade | |
Room Temperature |
Nucleic Acid Gel Electrophoresis & Blotting | |
Ambion™ | |
250 mL | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.