Learn More
Invitrogen™ Spin Cartridges
Replacement cartridges for the Invitrogen RNAqueous and the RNAqueous-4PCR kits.
Brand:  Invitrogen™ AM10051G
179.40 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 10176864
  • 276.00 € / Each
Explore more special offers
Description
Description
Replacement cartridges for the Invitrogen RNAqueous (Cat. No. AM1912) and the RNAqueous-4PCR (Cat. No. AM1914) kits.
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 cartridges; store at room temperature.
50 Each
Room Temperature