Learn More
Invitrogen™ T Cell Leukemia (Jurkat) Total RNA
Description
The highest-quality RNA available from Invitrogen, it is DNase-treated and subjected to unsurpassed quality control standards. One tube containing 100μg is provided at a concentration of 1mg/mL. Intactness of mRNA, genomic DNA contamination, presence/absence of enzymatic inhibitors, and ribosomal RNA contamination in poly(A) preparations are all important aspects used to judge the quality of FirstChoice™ RNAs. The samples are processed using Invitrogen RNA isolation reagents to produce highly pure, intact RNA. A stringent DNase treatment is performed to ensure that the RNA is ready for use in any downstream application, including RT-PCR. Because it is impossible to remove every DNA molecule from an RNA sample, each preparation is tested to ensure that any residual DNA contamination is insignificant. The integrity of the RNA is verified by capillary electrophoresis using the Agilent™ 2100 bioanalyzer.
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
| Cell Line | Jurkat |
| Concentration | 1 mg/mL |
| Content And Storage | Store at or below -70°C. Avoid multiple freeze–thaw cycles. Aliquots of the product may be stored short-term at -20°C. DO NOT STORE IN FROST-FREE FREEZER. |
| Species | Human |
| Sample Type | Total RNA |
| Product Line | Ambion |
| Quantity | 100 μg |
| Sample Treatment | DNase-Treated |
| Sample Source | Cell Line |
| Shipping Condition | Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.