missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TE, pH 7.0, RNase-free

Invitrogen TE buffer is a commonly recommended storage solution for RNA samples. 10 mM Tris-HCl, 1 mM EDTA, pH 7.0 is in nuclease-free, ultrapure water.

82.25 € - 83.25 €

Specifications

pH 7.0
Physical Form Liquid
Product Line Ambion
Packaging Bottle
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10546935
View Documents
Invitrogen™
AM9860
10 x 1 mL
83.25 €
10mL
Please to purchase this item. Need a web account? Register with us today!
10490975
View Documents
Invitrogen™
AM9861
50 mL
82.25 €
50mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Invitrogen TE buffer is a commonly recommended storage solution for RNA samples. 10 mM Tris-HCl, 1 mM EDTA, pH 7.0 is in nuclease-free, ultrapure water. The last step in every RNA isolation protocol is to resuspend the purified RNA pellet. After painstakingly isolating the RNA, it is crucial that the pellet be suspended and stored in a safe, RNase-free solution. TE Buffer is prepared from the purest reagents available, under scrupulously monitored conditions in a state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to guarantee their purity and performance.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

7.0
Liquid
Ambion
Bottle
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.