missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TE, pH 8.0, RNase-free

Invitrogen molecular biology grade, TE, pH 8.0 solution is supplied in one bottle containing 500 mL. The buffer is certified RNase-free, economical, and ready-to-use.

101.00 € - 122.00 €

Specifications

pH 8.0
Physical Form Liquid
Product Line Ambion
Packaging Bottle
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10742317
View Documents
Invitrogen™
AM9849
500 mL
101.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10006044
View Documents
Invitrogen™
AM9858
1 L
122.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Invitrogen molecular biology grade, TE, pH 8.0 solution is supplied in one bottle containing 500 mL. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

8.0
Liquid
Ambion
Bottle
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ TE, pH 8.0, RNase-free

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.