missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRI Reagent™ Solution

Tri Reagent™ solution is a complete, ready-to-use reagent for the isolation of total RNA or the simultaneous isolation of RNA, DNA, and protein from a variety of biological samples.

Brand:  Invitrogen™ AM9738


Product Code. 11312940

  • 255.00 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

100 mL TRI Reagent; 4°C

1 hr.
Bacteria, Cells, Plants, Tissue, Yeast
Not High-throughput Compatible (Manual)
100 mL
Bacteria: ≤107 cells
Cells: ≤106
Plant: ≤106 cells
Tissue: ≤100 mg
Yeast: ≤106 cells
Organic Extraction
Varies
Total RNA, DNA, Protein
RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern/Western blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization, enzymatic assays, immunoprecipitation, gel shift assays, 2D gel electrophoresis
Room Temperature
RNA: ≤15 μg (cells), ≤10 μg (tissue)
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2828146 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberAM9738
Lot Number2854904 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberAM9738
Lot Number2889917 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberAM9738
Lot Number2931350 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberAM9738
Lot Number2959369 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberAM9738
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ TRI Reagent™ Solution > 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.