missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIzol™ LS Reagent

TRIzol™ LS Reagent is a complete, ready-to-use reagent optimized for the isolation of high-quality total RNA, or the simultaneous isolation of RNA, DNA, and protein from a variety of liquid samples.

Brand:  Invitrogen™ 10296010

356.15 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.



 View more versions of this product

Product Code. 11578616

  • 419.00 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRIzol™ LS Reagent is a complete, ready-to-use reagent optimized for the isolation of high-quality total RNA, or the simultaneous isolation of RNA, DNA, and protein from a variety of liquid samples. This monophasic solution of phenol and guanidine isothiocyanate is designed to isolate separate fractions of RNA, DNA, and proteins from liquid samples of human, animal, plant, yeast, bacterial, and viral origin, typically within one hour.

Key features of TRIzol™ LS Reagent:
• Formulated for use with liquid samples such as serum and virus preparations
• Facilitates recovery of RNA, DNA, and protein from a single liquid sample
• Offers excellent lysis capability, even with difficult biological fluids

Reliably purify RNA from multiple sample sources
TRIzol™ LS Reagent is designed for processing a variety of liquid samples of up to 0.25 mL in volume. TRIzol™ LS Reagent differs from the standard TRIzol™ Reagent in concentration, which permits larger samples to be processed. Just as with the standard TRIzol™ Reagent, the integrity of resulting RNA preparations is maintained by the highly effective inhibition of RNase activity during sample homogenization. The simplicity of the TRIzol™ LS Reagent method allows simultaneous processing of a large number of samples. The entire procedure can be completed in 1 hour. Total RNA isolated by TRIzol™ LS Reagent is free of protein and DNA contamination.

Formulated to serve multiple isolations
TRIzol™ LS Reagent allows you to perform sequential precipitation of RNA, DNA, and proteins from a single sample. After homogenizing the sample with TRIzol™ LS Reagent, chloroform is added, and the homogenate is allowed to separate into a clear upper aqueous layer (containing RNA), an interface, and a red lower organic layer (containing the DNA and proteins). RNA is precipitated from the aqueous layer with isopropanol. DNA is precipitated from the aqueous/organic interface with ethanol. Protein is precipitated from the phenol-ethanol layer by isopropanol precipitation. The precipitated RNA, DNA, or protein is washed to remove impurities, and then resuspended for use in downstream applications.

Purified products are ideal for use with a variety of applications
Isolated RNA can be used in real-time quantitative PCR (qPCR), northern blot analysis, dot blot hybridization, poly-(A)+ selection, In vitro translation, RNase protection assays, and molecular cloning. Isolated DNA can be used in PCR, restriction enzyme digestion, and Southern blots. Isolated protein can be used for western blots, recovery of some enzymatic activity, and some immunoprecipitation.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mL TRIzol LS Reagent; room temperature

1 hr.
Liquid Samples (e.g. Serum)
Not High-throughput Compatible (Manual)
100 mL
0.25 mL per 0.75 mL of TRIzol™ LS Reagent
Organic Extraction
20 to 600 μL
Total RNA, DNA, Protein
RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern/Western blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization, enzymatic assays, immunoprecipitation, gel shift assays, 2D gel electrophoresis
Room Temperature
RNA: ≤20 μg per 1 mL blood
≤1.4 μg per 106 cells
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • TRIzol LS Reagent

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Thiocyanic acid, phenol, ammonium thiocyanate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.