Learn More
Invitrogen™ TURBO™ DNase (2 U/μL)
Efficiently degrades and digests DNA
204.00 € - 510.00 €
Specifications
Concentration | 2 U/μL |
---|---|
Content And Storage | TURBO™ DNase and 10X Reaction Buffer should be stored at –20°C. |
Shipping Condition | Dry Ice |
Enzyme | DNase |
Compatible Buffer | 10X Reaction Buffer |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10646175
|
Invitrogen™
AM2238 |
1000 U |
204.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10722687
|
Invitrogen™
AM2239 |
5000 U |
510.00 €
5000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cleaves double-stranded DNA nonspecifically to leave 5' phosphorylated oligodeoxynucleotides with an increased affinity for DNA-binding and remains active in presence of salt.
- Up to 50X more activity and 350% greater catalytic efficiency
- Efficiently degrades DNA in solutions containing up to 0.25M salt
- Efficiently digests DNA to oligonucleotides
- Used in clearing DNA templates from in vitro transcription reactions
- RNase-free and recombinant in origin
- Used to clear DNA contamination from RNA samples prior to RT-PCR
- Conventional DNase I has a poor affinity for DNA and cleaves DNA of low concentration
- Salt-sensitive; 20 mM NaCl can reduce activity of enzyme by 30%
- Purified from bovine pancreas, one of richest natural sources of RNase A
- Threat of contaminating RNase activity in DNase I preparations requires that enzyme be exhaustively purified
TURBO DNA-free (Mfr. No. AM1907) is available. It contains TURBO DNase along with reagents to inactivate the enzyme and remove divalent cations from the sample post-digestion.
PCR and Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription
Order Info
Shipping Conditions: Dry ice
Specifications
2 U/μL | |
Dry Ice | |
10X Reaction Buffer | |
Ambion™, TURBO™ |
TURBO™ DNase and 10X Reaction Buffer should be stored at –20°C. | |
DNase | |
DNase |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.