missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vortex Adapter-60

The adapter's unique shaking motion promotes rapid and thorough tissue digestion. Recommended for use with MELT™ Total RNA Isolation Kit (Cat. No. AM1983).

Brand:  Invitrogen™ AM10014

204.25 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 10361365

  • 222.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The adapter's unique shaking motion promotes rapid and thorough tissue digestion. Recommended for use with MELT™ Total RNA Isolation Kit (Cat. No. AM1983).

  • Vortex Adapter-60 consists of a 6 in. (152 mm) platform and two different Microplate Foam Inserts
  • Each insert fits snugly into platform to accommodate either 60 microtubes or one standard microplate
  • Compatible with Vortex-Genie™ 2, Vortex-Genie 2T, and VWR™ Mini Vortex Mixers

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature.
Room Temperature
1 each
Vortex
Ambion™
Tools and Accessories
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.