missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ AlkPhos Direct™ Labeling Modules

Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes. AlkPhos Direct Labeling Systems are based on the rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase.

801.00 € - 828.00 €

Specifications

Content And Storage 2°C to 8°C
For Use With (Application) Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes
Includes Labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer and blocking agent
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10298714
View Documents
Cytiva
RPN3680
2500 cm2 Membrane This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10074344
View Documents
Cytiva
RPN3681
For 5000 cm2 membrane This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

DNA or RNA probes are labeled during a 30-minute reaction and resulting probes can be used without purification. Hybridization stringency can be controlled by temperature as well as salt concentration.

  • Hybridization stringency can be controlled by elevating temperatures up to 75°C
  • Detection of 60 fg target DNA (single-copy gene) in a genomic Southern blot
  • Rapid protocol saves two to three hours over conventional indirect methods

Order Info

Each system include the following reagents, sufficient for 25 or 50 labeling reactions and detection of 2500cm2 or 5000cm2 membrane (depending on product ordered): labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer and blocking agent

Specifications

Specifications

2°C to 8°C
Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes
Labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer and blocking agent
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : methanol, formaldehyde
  • UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.