Learn More
Cytiva Amersham™ CDP-Star Detection Reagents
Rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase. Cytiva Amersham™ CDP-Star Detection Reagents label DNA or RNA probes during a 30-minute reaction and resulting probes can be used without purification.
Brand: Cytiva RPN3682
568.10 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
Description
- Convenient for rapid separations either alone or connected in series
- Designed for use with syringe, peristaltic pump or chromatographic systems
- High flowrates and good scale-up potential
Order Info
Sufficient for detection of 2500cm2 membrane. Includes detection reagents only. For labeling module, see part RPN3680.
Specifications
DNA/RNA and Probes Labeling | |
2500cm2 membrane | |
CDP-Star detection reagents | |
Each |
Safety and Handling
- Danger||
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
- P305-IF IN EYES:Immediately call a POISON CENTER/doctor..
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 2,2'-iminodiethanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.