missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Cy™3, Cy™ 5 Maleimide mono-Reactive Dye 5-packs

More selective method of antibody labeling that reduces the possibility of the label interfering with subsequent antibody-antigen reactions. Amersham™ Cy™3, Cy™ 5 Maleimide mono-Reactive Dye packs are packaged in premeasured amounts and foil-sealed to ensure consistent labelings.

Brand:  Cytiva PA25031

338.17 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 10704357

  • 380.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Cy3 and Cy5 mono-functional maleimides are particularly suitable for the selective labeling of molecules containing free sulfhydryl groups, such as cysteine residues in proteins and peptides, and oligonucleotides. They are also useful in antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired. The protocol is simple and robust, offering an alternative chemistry when either greater selectivity of labeling is required or amine labeling with Cy3 and Cy5 NHS esters is not desirable or appropriate.

  • Provides an alternative for when CyDye™ NHS esters are unsuitable.
  • Dyes are packaged in premeasured amounts and foil-sealed to ensure consistent labelings

Order Info

Each kit contains five vials of dye, each sufficient for labeling 1mg of protein.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

For the labelling of biological compounds with CyTM5 monofunctional dye
Cy5 Maleimide Mono-Reactive Dye 5-Pack
Labeling 1mg Protein
Soluble in cold water and hot water
Cy5
5 vials each sufficient to label 1 mg protein
Five vials of dye
817
2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
Blue
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1

SDS
Documents

Documents

Certificates
Special Offers

Special Offers