missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Cy™ 3 Saturation Dye
Outstanding for multi-color analysis, with bright and easily distinguishable colors
Brand: Cytiva 28-9366-83
Includes: 400nmol CyDye DIGE fluor Cy3 saturation dye
Description
- Water soluble at low and high temperatures
- Bright coloration, very sensitive
- Minimal loss of signal during labeling, separation and scanning
- Distinct signal from each fluor with minimal cross-talk
- No change in signal over wide pH range used during first-dimension separation
Detecting protein abundance differences in 2-D Fluorescence Difference Gel Electrophoresis using Ettan DIGE system; For use with very small samples
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Red | |
Fluorescent Protein Stain | |
Solid | |
400 nmol CyDye DIGE fluor Cy3 saturation dye for preparative labeling | |
−15° to −30°C |
400 nmol | |
CyDye DIGE Fluor | |
Multicolor analysis, offering bright and intense colors with narrow excitation and emission bands | |
400 nmol |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction