missing translation for 'onlineSavingsMsg'
Learn More

Amersham™ CyDye™ 700 Goat-Anti-Rabbit Secondary Antibody, Cytiva

Amersham™ CyDye™ NIR-labeled secondary antibodies are designed for quantitative fluorescent Western blotting.

123.98 € - 211.00 €

Specifications

Applications Western Blot
Conjugate CyDye 700
Primary or Secondary Secondary
Content And Storage 2°C to 8°C, protected from light
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16217801
View Documents
Life Science products Cytiva
29360786
0.1 mg
123.98 €
0.10mg
Please to purchase this item. Need a web account? Register with us today!
16227801
View Documents
Life Science products Cytiva
29360787
0.5 mg
211.00 €
0.50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fluorescence detection is a direct Western blotting method in which a secondary antibody is conjugated to a fluorophore, avoiding the need for ancillary detection reagents. Different fluorophores are available for signal detection at either visible light or near-infrared (NIR) wavelengths.

Amersham CyDye 700 and 800 secondary antibodies are labeled with NIR fluorophores that emit light at wavelengths of 700 nm or 800 nm. By using Amersham CyDye 700 and 800 secondary antibodies together with a pair of appropriate rabbit or mouse primary antibodies, you can perform multiplexed experiments.

Key benefits:

  • Easy handling and a strong signal-to-noise ratio yield reproducible results
  • Multiplexing analysis of more than one target protein on a single blot increases process efficiency
  • Total protein normalization (TPN) generates reliable and accurate quantitation
  • Stable signals allow membranes to be stored and rescanned for future studies
Specifications

Specifications

Western Blot
CyDye 700
Secondary
2°C to 8°C, protected from light
Videos
Safety and Handling

Safety and Handling

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.