missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ CyDye™ Antibody Labeling Kits
Convenient way to tag antibodies with CyDye fluorescent dyes
172.00 € - 629.00 €
Specifications
Quantity | Each |
---|---|
pH Range | 7 to 7.3 |
Content And Storage | 2°C to 8°C, protected from light |
Product Code | Brand | For Use With (Application) | Label or Dye | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Label or Dye | Product Type | Price | Quantity & Availability | |||
10744167
|
Cytiva
PA32000 |
For the conjugation of proteins with Cy2 Reactive Dye | Cy2 Reactive Dye | Cy2 Ab Labeling Kit | N/A | ||||
10106334
|
Cytiva
PA35000 |
For the conjugation of proteins with Cy5 Reactive Dye | Cy5 Reactive Dye | Cy5 Ab Labeling Kit |
581.00 €
1 set |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||
10754167
|
Cytiva
PA33000 |
For the conjugation of proteins with Cy3 Reactive Dye | Cy3 Reactive Dye | Cy3 Ab Labeling Kit |
629.00 €
1 set |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||
10685375
|
Cytiva
PA32001 |
For the conjugation of proteins with Cy2 bisfunctional dye | Cy2 mono-Functional Dye | Cy2 MAb Labeling Kit | N/A | ||||
Description
CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
- Conjugations are carried out under mild conditions and are complete within 1 hour
- Dyes are packaged in premeasured amounts and foil sealed to ensure consistent labelings
Order Info
Antibody kits contain all reagents required for 2 x 1mg reactions. Monoclonal antibody kits contain all reagents required for 2 x 0.1mg reactions.
Specifications
Each | |
7 to 7.3 | |
2°C to 8°C, protected from light |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1