Learn More
Cytiva Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System
Use for fast and easy detection of medium-to-high-target amounts. Cytiva Life Sciences™ Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 min chemical reaction
556.00 € - 2024.00 €
Includes: 45-000-922 and 45-000-923: Labeling reagent; Crosslinker; Control DNA; Blocking agent; ECL detection reagents and ECL Gold hybridization buffer
Products 6
Product Code Brand Includes Quantity Price Quantity & Availability  
Product Code Brand Includes Quantity Price Quantity & Availability  
10733797
View Documents
Cytiva
RPN3005
DNA labelling reagent 0.5mL; Glutaraldehyde solution 0.5mL; Water 1.0mL; l Hind III control DNA (10μg/mL) 0.1 ml; Gold hybridization buffer 500mL; Blocking agent 25g To LabeL 5 μg
799.00 €
5µg
10195804
View Documents
Cytiva
RPN2105
ECL Detection Reagents 1 and 2, for 4000cm2 membrane For 4000 cm2 membrane
772.00 €
Each
10554205
View Documents
Cytiva
RPN3001
DNA labeling reagent (2 x 0.5mL); Glutaraldehyde solution (2 x 0.5mL); Water (2 x 1.0mL); l Hind III control DNA 10μg/mL (2 x 0.1mL); Gold hybridization buffer (2 x 500mL); Blocking agent (2 x 25g); ECL detection reagent 1 (2 x 125mL); ECL detection reagent 2 (2 x 125mL) For Labeling 10μg Nucleic Acid
2024.00 €
Each
10411095
View Documents
Cytiva
RPN3004
ECL Detection Reagents 1 and 2, for 2000cm2 membrane For 2000cm2 Membrane
556.00 €
Each
10269004
View Documents
Cytiva
RPN3000
DNA labeling reagent 0.5mL; Glutaraldehyde solution 0.5mL; Water 1.0mL; l Hind III control DNA (10μg/mL) 0.1mL; Gold hybridization buffer 500mL; Blocking agent 25g; ECL detection reagent 1 125mL; ECL detection reagent 2 125mL Labels 5 μg Nucleic Acid
1079.00 €
Each
10459774
View Documents
Cytiva
RPN3006
ECL Gold Hybridization Buffer sufficient for 4000 cm² Sufficient for 4000 cm2
1628.00 €
Each
Description
Description
    • Direct probe labeling in a 10 min reaction, 1 h from hybridization to detection with ECL Direct, Hybond N+, and Hyperfilm ECL.
    • Eliminates handling, waste, and regulatory issues associated with the use of radioactivity.
    • No need to strip blots before reprobing.
    • For fast and easy detection of medium- to high-target amounts in applications such as colony/plaque screens, dot blots, and PCR product analyses.
    • Consistent results combining strong signals with very low backgrounds.
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 3 Months
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P405-Store locked up.
  • P501-Dispose of contents/container to .