missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL™ start Western Blotting Detection Reagent

Reagent is luminol-based chemiluminescent substrate designed for the detection of proteins immobilized to a membrane

Brand:  Cytiva RPN3244

196.97 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 15397655

  • 241.00 € / 400mL
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Amersham™ ECL™ start is a chemiluminescent detection reagent in the Amersham ECL product family that is recommended for confirmatory analysis and detection of high to medium levels of endogenous proteins.

  • Long signal duration provides assay design flexibility and robust detection of large experimental sets
  • Produces low background, regardless of method of detection
  • Working solution is stable for five days
  • Long shelf life of 18 hours is convenient for those who run Western blots infrequently
  • Detection on X-ray film or CCD Imager
  • Recommended Membrane: PVDF or nitrocellulose
  • For detection of high abundant proteins
  • Colorless detection reagent 1 and 2

Applications:

Routine and confirmatory Western blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ECL Western Blotting Detection Reagent
400 mL
Routine and confirmatory Western blotting
18°C to 30°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Statement
  • EUH210-Safety data sheet available on request.

Supplemental information
  • UNKNOWN ACUTE-4 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 4% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers