Learn More
Cytiva Amersham™ ECL™ Western Blotting Reagent Pack
Reagents for membrane blocking and secondary detection of membrane-bound mouse and rabbit primary antibodies
Brand: Cytiva RPN2124
Includes: anti-mouse IgG horseradish peroxidase conjugate, 100μL; anti-rabbit IgG horseradish peroxidase conjugate, 100μL; Amersham ECL blocking agent, 5g
Description
- Designed for use with nitrocellulose or PVDF membranes
- System optimized for use with ECL detection reagents (Not included)
Specifications
High sensitivity western blotting and chemifluorescent detection on scanners | |
100μL Anti-Mouse IgG, Peroxidase linked whole Antibody (from Sheep), 100μL Anti-Rabbit IgG, Peroxidase linked whole Antibody (from Donkey), 5g ECL™ membrane blocking agent | |
Each |