missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Hybond™ Membranes

Unsupported 100% pure nitrocellulose membrane validated for use with all ECL Western and protein blotting applications

Brand:  Cytiva RPN78D

 View more versions of this product

Product Code. 11738388

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Excellent sensitivity, resolution and low background for all labeling and detection systems: radioactive, nonradioactive and chromogenic
  • Packaging features include interleaf on both sides of the membrane and airtight resealable bag with marking spot
  • Pore size: 0.2μm and 0.45μm
  • Discs are also available

TRUSTED_SUSTAINABILITY
Specifications

Specifications

7 cm
7 x 8cm
50 Sheets
Nitrocellulose
8 cm
Protein blotting
Sheet
0.45 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.