Learn More
Cytiva Amersham™ Hybond™ P 0.45μm PVDF Membrane
Use for chemiluminescent and fluorescent detection methods for proteins of > Mr 20 000. Cytiva Life Sciences™ Amersham™ Hybond™ P 0.45μm PVDF Membrane is a 0.45 μm pore size hydrophobic membrane, with high physical strength, highly suitable for stripping and reprobing.
133.78 € - 443.00 €
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
15289894
|
Cytiva
10600069 |
140 x 160mm | 25 Sheets/Pk. |
281.00 €
25 sheets |
In Stock
Log in to see stock. |
||||
15259894
|
Cytiva
10600023 |
300mm x 4m | 1 Roll/Pk. |
414.00 €
1 roll |
In Stock
Log in to see stock. |
||||
15279894
|
Cytiva
10600038 |
300 x 600mm | 5/Pk. |
443.00 €
5 sheets |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||||
15269894
|
Cytiva
10600029 |
150mm x 4m | 1 Roll/Pk. |
269.00 €
1 roll |
In Stock
Log in to see stock. |
||||
15209904
|
Cytiva
10600087 |
100 x 100mm | 10 Sheets/Pk. |
133.78 €
10 sheets |
In Stock
Log in to see stock. |
||||
15299894
|
Cytiva
10600086 |
100 x 100mm | 25 Sheets/Pk. |
210.00 €
25 sheets |
In Stock
Log in to see stock. |
||||
15219904
|
Cytiva
10600100 |
80 x 90mm | 25 Sheets/Pk. |
144.22 €
25 sheets |
In Stock
Log in to see stock. |
||||
Description
- Protein binding capacity over over 200 μg/cm2 for easy signal detection
- High protein retention even after harsh washing steps
- High protein capture during transfer minimizes sample loss
- Available in popular precut sizes for your application
Specifications
0.45 μm |