missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Hybond™ PVDF Membranes: Rolls

Ideal for Western blotting applications where stripping and reprobing are necessary. Cytiva Life Sciences™ Amersham™ Hybond™ PVDF Membranes: Rolls are hydrophobic PVDF membranes.

Brand:  Cytiva 10600022

547.57 EUR valid until 2024-12-31
Use promo code "21973" to get your promotional price.


 View more versions of this product

Product Code. 15279884

  • 671.00 € / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Ideal for Western blotting applications where stripping and reprobing are necessary. Cytiva Life Sciences™ Amersham™ Hybond™ PVDF Membranes: Rolls are hydrophobic PVDF membranes.

  • Supplied in rolls.
  • Hydrophobic PVDF membranes
  • Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights

Amersham Hybond 0.2 μm

  • Compatible with a range of solvents used for rapid destaining

Amersham Hybond SEQ 0.2 μm

  • Specifically designed for protein sequencing applications and solid phase amino acid analysis
  • Offers excellent blotting for proteins Mr<20,000

Amersham Hybond LFP 0.2 μm

  • Specifically optimized for fluorescence detection in Western blotting applications
  • Provides low background fluorescence, resulting in high sensitivity
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Specifically optimized for fluorescence detection in Western blotting applications
Roll
4 m x 260 mm
LFP 0.2 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers