missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Hybond™ PVDF Membranes: Sheets

Ideal for Western blotting applications where stripping and reprobing are necessary

Brand:  Cytiva 10600088

 View more versions of this product

Product Code. 15219864

This item is not returnable. View return policy

Description

Description

Individual sheets.

  • Hydrophobic PVDF membranes
  • Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights

Amersham Hybond 0.2μm

  • Compatible with a range of solvents used for rapid destaining
  • Amersham Hybond SEQ 0.2μm
    • Specifically designed for protein sequencing applications and solid phase amino acid analysis
    • Offers excellent blotting for proteins Mr<20,000
    Amersham Hybond LFP 0.2μm
    • Specifically optimized for fluorescence detection in Western blotting applications
    • Provides low background fluorescence, resulting in high sensitivity

    TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    Hybond P
    25/Pk.
    PVDF
    Ideal for Western blotting applications where stripping and reprobing are necessary
    Sheet
    10 x 10 cm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers