Learn More
Cytiva Amersham™ Megaprime™ DNA Labeling System
For labeling DNA probes to specific activities >1 x 109dpm/μg in 10 minutes
Brand:  Cytiva RPN1607
 View more versions of this product
Product Code. 10175804
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes: Primer solution; Labeling buffer; Reaction buffer; Enzyme; Standard DNA solution; Carrier DNA solution
Description
Description
Megaprime™ DNA Labeling Systems are used for rapid, random-prime labeling from as little as 25ng substrate DNA.
  • Features cloned Klenow fragment and random nonamers in the random-prime labeling technique
  • No need to remove unincorporated nucleotides before hybridization
  • Protocols for labeling directly from low melting point agarose
TRUSTED_SUSTAINABILITY
Specifications
Specifications
-15°C to -30°C
Primer solution 300μL, Labeling Buffered ATP, dGTP and dTTP in Tris HCl pH7.5, 2-mercaptoethanol and MgCl2 600μL, Nucleotide solutions dATP, cCTP, dGTP, dTTP in Tris HCl pH 8, 0.5mM EDTA, Reaction Buffer A 10 x concentr
Megaprime DNA Labeling System, dCTP (for use with labeled dCTP)
Membrane hybridization
60