Learn More
Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls
Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
243.55 € - 508.00 €
Specifications
Length (Metric) | 4 m |
---|---|
For Use With (Application) | Western blots, dot-blot assays, and other protein or nucleic acid methods |
Format | Roll |
Material | 100% Pure Nitrocellulose |
Product Code | Brand | Width (Metric) | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Pore Size | Price | Quantity & Availability | ||||
15239794
|
Cytiva
10600000 |
30 cm | 0.1 μm |
508.00 €
1 roll |
In Stock
Log in to see stock. |
||||
15259794
|
Cytiva
10600002 |
30 cm | 0.45 μm |
370.00 €
1 roll |
In Stock
Log in to see stock. |
||||
15249794
|
Cytiva
10600001 |
30 cm | 0.2 μm |
447.00 €
1 roll |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15289794
|
Cytiva
10600005 |
20 cm | 0.1 μm |
388.00 €
1 roll |
In Stock
Log in to see stock. |
||||
15299794
|
Cytiva
10600006 |
20 cm | 0.2 μm |
323.00 €
1 roll |
In Stock
Log in to see stock. |
||||
15239804
|
Cytiva
10600010 |
15 cm | 0.1 μm |
326.00 €
1 roll |
In Stock
Log in to see stock. |
||||
15259804
|
Cytiva
10600012 |
15 cm | 0.45 μm |
243.55 €
1 roll |
In Stock
Log in to see stock. |
||||
15209804
|
Cytiva
10600007 |
20 cm | 0.45 μm |
297.00 €
1 roll |
In Stock
Log in to see stock. |
||||
15249804
|
Cytiva
10600011 |
15 cm | 0.2 μm |
265.00 €
1 roll |
In Stock
Log in to see stock. |
||||
Description
Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Packed in rolls.
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
- Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Specifications
4 m | |
Western blots, dot-blot assays, and other protein or nucleic acid methods | |
Roll | |
100% Pure Nitrocellulose |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.