missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets

Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

119.14 € - 547.00 €

Specifications

For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Format Sheet
View More Specs

Products 15
Product Code Brand Length (Metric) Width (Metric) Pore Size Quantity Price Quantity & Availability  
Product Code Brand Length (Metric) Width (Metric) Pore Size Quantity Price Quantity & Availability  
15279814
View Documents
Cytiva
10600033
60 cm 30 cm 0.45 μm 5/Pk.
427.00 €
5 sheets
In Stock
to see stock.
Add to basket
 
15269814
View Documents
Cytiva
10600032
60 cm 30 cm 0.2 μm 5/Pk.
457.00 €
5 sheets
In Stock
to see stock.
Add to basket
 
15239824
View Documents
Cytiva
10600041
20 cm 20 cm 0.45 μm 25/Pk.
419.00 €
25 sheets
In Stock
to see stock.
Edge
Add to basket
 
15249824
View Documents
Cytiva
10600042
20 cm 20 cm 0.45 μm 10/Pk.
283.00 €
10 sheets
In Stock
to see stock.
Add to basket
 
15259824
View Documents
Cytiva
10600043
20 cm 20 cm 0.2 μm 25/Pk.
443.00 €
25 sheets
In Stock
to see stock.
Add to basket
 
15269824
View Documents
Cytiva
10600044
20 cm 20 cm 0.2 μm 10/Pk.
274.00 €
10 sheets
In Stock
to see stock.
Add to basket
 
15279824
View Documents
Cytiva
10600045
20 cm 20 cm 0.1 μm 25/Pk.
547.00 €
25 sheets
In Stock
to see stock.
Add to basket
 
15289824
View Documents
Cytiva
10600046
20 cm 20 cm 0.1 μm 10/Pk.
332.00 €
10 sheets
In Stock
to see stock.
Add to basket
 
15209844
View Documents
Cytiva
10600062
16 cm 14 cm 0.45 μm 25/Pk.
255.00 €
25 sheets
In Stock
to see stock.
Add to basket
 
15289844
View Documents
Cytiva
10600073
10 cm 10 cm 0.45 μm 10/Pk.
119.14 €
10 sheets
In Stock
to see stock.
Add to basket
 
15299844
View Documents
Cytiva
10600074
10 cm 10 cm 0.2 μm 25/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15249864
View Documents
Cytiva
10600093
8 cm 9 cm 0.45 μm 25/Pk.
131.58 €
25 sheets
In Stock
to see stock.
Add to basket
 
15269864
View Documents
Cytiva
10600095
8 cm 9 cm 0.1 μm 25/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15259864
View Documents
Cytiva
10600094
8 cm 9 cm 0.2 μm 25/Pk.
124.48 €
25 sheets
In Stock
to see stock.
Add to basket
 
15265928
View Documents
Cytiva
10600125
10.2 cm 13.3 cm 0.2 μm 10/Pk.
139.19 €
10 sheets
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

Individual sheets.

  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent

Specifications

Specifications

Western blots, dot-blot assays, and other protein or nucleic acid methods
Sheet
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.