Learn More
Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets
Use for high sensitivity, resolution, and low background for all labeling and detection systems, including chemiluminescent, fluorescenct, colorimetric and radioactive. Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
106.29 € - 452.00 €
Specifications
For Use With (Application) | Western blots, dot-blot assays, and other protein or nucleic acid methods |
---|---|
Format | Sheet |
Product Code | Brand | Length (Metric) | Width (Metric) | Pore Size | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Pore Size | Quantity | Price | Quantity & Availability | ||
15289814
|
Cytiva
10600034 |
60 cm | 30 cm | 0.45 μm | 5/Pk. |
435.00 €
5 sheets |
In Stock
Log in to see stock. |
||
15299824
|
Cytiva
10600047 |
20 cm | 20 cm | 0.45 μm | 25/Pk. |
452.00 €
25 sheets |
In Stock
Log in to see stock. |
||
15209834
|
Cytiva
10600048 |
20 cm | 20 cm | 0.45 μm | 10/Pk. |
252.00 €
10 sheets |
In Stock
Log in to see stock. |
||
15239844
|
Cytiva
10600065 |
16 cm | 14 cm | 0.45 μm | 25/Pk. |
257.00 €
25 sheets |
In Stock
Log in to see stock. |
||
15249854
|
Cytiva
10600079 |
10 cm | 10 cm | 0.45 μm | 10/Pk. |
123.52 €
10 sheets |
In Stock
Log in to see stock. |
||
15259854
|
Cytiva
10600080 |
10 cm | 10 cm | 0.2 μm | 25/Pk. |
233.00 €
25 sheets |
In Stock
Log in to see stock. |
||
15269854
|
Cytiva
10600081 |
10 cm | 10 cm | 0.2 μm | 10/Pk. | N/A | |||
15289864
|
Cytiva
10600097 |
9 cm | 8 cm | 0.2 μm | 25/Pk. | N/A | |||
15279864
|
Cytiva
10600096 |
9 cm | 8 cm | 0.45 μm | 25/Pk. |
133.17 €
25 sheets |
In Stock
Log in to see stock. |
||
Description
Use for high sensitivity, resolution, and low background for all labeling and detection systems, including chemiluminescent, fluorescenct, colorimetric and radioactive. Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Individual sheets.
- Low background enables a dynamic range for complicated samples
- Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
Specifications
Western blots, dot-blot assays, and other protein or nucleic acid methods | |
Sheet |
Safety and Handling
- Amersham Protran Premium
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.