Learn More
Cytiva Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Rolls
Use for robust protein blotting. Cytiva Life Sciences™ Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Brand: Cytiva 10600016
352.53 EUR valid until 2025-12-31
Use promo code "24363" to get your promotional price.
Description
Use for robust protein blotting. Cytiva Life Sciences™ Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Roll dispensers.
- Membranes consist of nitrocellulose supported by an inert polyester nonwoven material within the membrane
- The support does not affect transfer conditions or results
Specifications
4 m | |
Each | |
Nitrocellulose, Inert Polyester | |
4 m x 30 cm |
30 cm | |
Roll | |
0.45 μm | |
Amersham Protran |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : Nitrocellulose
Your input is important to us. Please complete this form to provide feedback related to the content on this product.