missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Rapid-hyb Buffer

Pre-mixed, rate-enhancing hybridization buffer. Amersham™ Rapid-hyb Buffer reduces hybridization time for detecting single-copy mammalian genes to just two hours.

242.00 €

Specifications

Content And Storage Store at ambient temperature. Under these conditions the product is stable for at least 3 months
For Use With (Application) Radiolabeled DNA, RNA or Oligonucleotide probes
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11585055
View Documents
Cytiva
RPN1636
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • For use with radiolabeled DNA, RNA, or oligonucleotide probes
  • Pre-mixed solution improves reproducibility between experiments
  • Suited for Southern and Northern blotting applications
  • Available in two different quantities, sufficient for 1000cm2 membrane and 4000cm2 membrane

Developed for use with nylon membranes; not suitable for use with PVDF or nitrocellulose membranes.

Specifications

Specifications

Store at ambient temperature. Under these conditions the product is stable for at least 3 months
Radiolabeled DNA, RNA or Oligonucleotide probes
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Rapid-hyb Buffer sufficient for 4,000 cm2 membrane

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.

Supplemental information
  • MIXTURE LIST-Contains : 2-hydroxy-1,2,3-Propanetricarboxylic acid hydrate
  • UNKNOWN ACUTE-19 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 6% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Product Certifications