missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Streptavidin-Fluor Conjugates
Use for in situ hybridization and flow cytometry. Cytiva Life Sciences™ Amersham™ Streptavidin-Fluor Conjugates consist of streptavidin conjugated to a range of fluorescent dyes.
240.00 € - 449.00 €
Specifications
Formulation | This product is supplied as a lyophilized solid in Phosphate-buffered Saline (0.02M Potassium Phosphate, 0.15M NaCl) pH 7.2, containing 1%(w/v) Bovine Serum Albumin and 0.1% (w/v) Sodium Azide, reconstitute with 1mL of deionized Water, dispense into suitable aliquots and freeze, avoid repeated freeze-thaw cycles |
---|---|
For Use With (Application) | Suitable for use in the Fluorescent analysis of Biotinylated samples |
Quantity | 1 mg |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
10544765
|
Cytiva
PA43001 |
Cy3-Streptavidin |
240.00 €
Each |
In Stock
Log in to see stock. |
|||||
10277704
|
Cytiva
PA42001 |
Cy2-Streptavidin | N/A | ||||||
10004594
|
Cytiva
PA45001 |
Cy5-Streptavidin |
295.00 €
1mg |
In Stock
Log in to see stock. |
|||||
Description
- High specificity reagents
Applications:
in situ hybridization, flow cytometry
Specifications
This product is supplied as a lyophilized solid in Phosphate-buffered Saline (0.02M Potassium Phosphate, 0.15M NaCl) pH 7.2, containing 1%(w/v) Bovine Serum Albumin and 0.1% (w/v) Sodium Azide, reconstitute with 1mL of deionized Water, dispense into suitable aliquots and freeze, avoid repeated freeze-thaw cycles | |
Suitable for use in the Fluorescent analysis of Biotinylated samples | |
1 mg |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501-Dispose of contents/container to .
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title