missing translation for 'onlineSavingsMsg'
Learn More

Merck Amicon™ Stirred Cell Seal Kits

Use with solutions up to 10% macrosolute concentration. Amicon™ Stirred Cell Seal Kit includes 3 silicone gaskets and O-rings. The Amicon stirred cell model provides high flow rates and is capable of rapid concentration or salt removal / concentration in the same unit.

Brand:  Merck UFSC400SL

 View more versions of this product

Product Code. 15897364

  • 235.00 € / 400mL
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Amicon™ Stirred Cell Seal Kit (3 each o-ring and gasket)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
41.8 cm2
Amicon™ Stirred Cell 400 mL Seal Kit (3 Ea. of o-ring and gasket)
Ultrafiltration Equipment
18.7 cm
76 mm (filter)
Silicone
Protein desalting, purification and concentration
3 Ea. gaskets and O-rings
400 mL
76 mm filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.