missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Amicon™ Stirred Cells
Amicon stirred cells concentrate samples rapidly but gently, using magnetic stirring to minimize concentration polarization and shear stress-induced denaturation.
2340.00 € - 3945.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
For Use With (Equipment) | Ultracel™, Biomax™, and Durapore™ membrane discs for use in stirred cells |
For Use With (Application) | Concentration, Buffer Exchange, Desalting |
Type | Ultrafiltration Equipment |
Alert:
Pressure Limitation. Do not operate the stirred cell above 5.2 bar (75 psi).
Product Code | Brand | Volume (Metric) Hold Up | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) Hold Up | Price | Quantity & Availability | ||||||
15572061
|
Merck
UFSC05001 |
50 mL |
2340.00 €
Each |
Estimated Shipment: 13-06-2025
Log in to see stock. |
||||||
15970817
|
Merck
UFSC20001 |
200 mL |
3045.00 €
Each |
Estimated Shipment: 13-06-2025
Log in to see stock. |
||||||
15573333
|
Merck
UFSC40001 |
400 mL |
3945.00 €
Each |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
Description
This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.Specifications
Autoclavable | |
Ultracel™, Biomax™, and Durapore™ membrane discs for use in stirred cells | |
Concentration, Buffer Exchange, Desalting | |
Ultrafiltration Equipment |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Merck Amicon™ Stirred Cells