missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amikacin (From Kanamycin A, White Crystalline Powder), Fisher BioReagents
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass
Description
- An antibiotic that serves as a selective agent to isolate transformed bacteria
- Amikacin functions as an indirect inhibitor of protein synthesis in prokaryotes by binding to the 30S subunit of ribosomes
- Useful as a selective agent to isolate bacteria that have been transformed with plasmid containing the antibiotic-resistant gene
- Intended for research purposes only; not for manufacturing or commercial use
Specifications
Specifications
Type | Amikacin |
Physical Form | Solid |
pH | 9.5 to 11.5 |
Melting Point | 214°C |
Merck Index | 15, 400 |
Packaging | Amber Glass |
Potency | > 900μg/mg |
Solubility Information | completely soluble in water |
Formula Weight | 585.61 |
Color | White |
Show More |
Safety and Handling
Product Identifier
- Amikacin
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction