Learn More
Novus Biologicals™ Amine Reactive Comp-Bead 2 Population Kit
The Amine Reactive Compensation Beads are designed for labeling amine-reactive dyes (live/dead fixable viability stains) when setting compensation in flow cytometry.
Brand:  Novus Biologicals™ NBP3-00496
Product Code. 18075196
  • 455.95 € / 1 set
Explore more special offers
Specifications
Store at 4°C in the dark. Do not freeze.
10 mL (2 x 5 mL)
0.016 M PBS (pH 7.4)
Flow Cytometry