missing translation for 'onlineSavingsMsg'
Learn More

(Aminoethylaminomethyl)phenethyltrimethoxysilane, mixture of m and p isomers, Thermo Scientific Chemicals

Product Code. 11344948
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

As a crosslinking agent

As a crosslinking agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74113-77-2
Molecular Formula H2NC2H4NHCH2C6H4C2H4Si(OCH3)3
MDL Number MFCD00054775
Synonym aminoethy-laminomethyl phenethyl-trimethoxysilane, 4-2-aminoethylaminomethyl phenethyltrimethoxysilane, n1-4-2-trimethoxysilyl ethyl benzyl ethane-1,2-diamine, n'-4-2-trimethoxysilylethyl phenyl methyl ethane-1,2-diamine, 2-aminoethyl 4-2-trimethoxysilyl ethyl phenyl methyl amine

Specifications

Boiling Point 128°C (0.2mmHg)
Color Colorless to Yellow
Physical Form Liquid
Quantity 50 g
UN Number UN2735
Flash Point >110°C (230°F)
Formula Weight 298.46
Sensitivity Air & Moisture Sensitive
Density 1.02g/mL
Chemical Name or Material (Aminoethylaminomethyl)phenethyltrimethoxysilane, Mixture of m and p isomers

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'