missing translation for 'onlineSavingsMsg'
Learn More

Aminomalononitrile p-toluenesulfonate, 97+%, Thermo Scientific Chemicals

Product Code. 11352826
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5098-14-6
Molecular Formula C10H11N3O3S
Molecular Weight (g/mol) 253.276
MDL Number MFCD00012848
InChI Key MEUWQVWJLLBVQI-UHFFFAOYSA-N
Synonym aminomalononitrile p-toluenesulfonate, 2-aminomalononitrile 4-methylbenzenesulfonate, aminomalononitrile p-toluenesulphonate, aminomalononitrile p-toluenesulfonic acid, aminomalononitrile tosylate, aminomalononitrile-p-toluene sulfonate, 2-aminomalononitrile 4-methylbenzenesulphonate, 2-aminomalononitrile p-toluenesulfonate, acmc-209krb, dicyanomethanaminium 4-methylbenzenesulfonate
PubChem CID 563049
IUPAC Name 2-aminopropanedinitrile;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C(#N)C(C#N)N

Specifications

Melting Point ∽169°C (decomposition)
Quantity 5 g
UN Number UN3439
Beilstein 4050747
Formula Weight 253.28
Percent Purity ≥97%
Chemical Name or Material Aminomalononitrile p-toluenesulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.