Learn More
(Aminomethyl)cyclopropane, 97%, Thermo Scientific Chemicals
85.50 € - 638.00 €
Chemical Identifiers
CAS | 2516-47-4 |
---|---|
Molecular Formula | C4H9N |
Molecular Weight (g/mol) | 71.11 |
InChI Key | IGSKHXTUVXSOMB-UHFFFAOYSA-N |
Synonym | cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine |
PubChem CID | 75646 |
IUPAC Name | cyclopropylmethanamine |
SMILES | C1CC1CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10622983
|
Thermo Scientific Acros
258720050 |
5 g | Glass bottle |
85.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10666271
|
Thermo Scientific Acros
258720100 |
10 g | Glass bottle |
156.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11915201
|
Thermo Scientific Acros
258720500 |
50 g | Glass bottle |
638.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2516-47-4 | |
71.11 | |
cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine | |
cyclopropylmethanamine |
C4H9N | |
IGSKHXTUVXSOMB-UHFFFAOYSA-N | |
75646 | |
C1CC1CN |
Specifications
2516-47-4 | |
0.8200g/mL | |
−30°C | |
96% min. (GC) | |
1.4320 to 1.4340 | |
cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine | |
C1CC1CN | |
71.11 | |
71.11 | |
Liquid |
Colorless to Yellow | |
84.0°C to 85.0°C | |
Authentic | |
C4H9N | |
0.82 | |
IGSKHXTUVXSOMB-UHFFFAOYSA-N | |
cyclopropylmethanamine | |
75646 | |
97% | |
(Aminomethyl)cyclopropane |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 219-737-6
RUO – Research Use Only