missing translation for 'onlineSavingsMsg'
Learn More

(Aminomethyl)cyclopropane hydrochloride, 97%, Thermo Scientific™

Product Code. 11905201
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7252-53-1
Molecular Formula C4H9ClN
Molecular Weight (g/mol) 106.57
MDL Number MFCD00012544
InChI Key MQCZBGGAZQXTLH-UHFFFAOYSA-N
Synonym cyclopropanemethylamine hydrochloride, cyclopropylmethanamine hydrochloride, cyclopropylmethyl ammonium chloride, cyclopropylmethylamine hydrochloride, aminomethylcyclopropane hydrochloride, 1-cyclopropylmethanamine hydrochloride, aminomethyl cyclopropane hydrochloride, cyclopropylmethanaminium chloride, cyclopropylmethylamin hydrochloride, cyclopropylmethanamine-hydrochloride
PubChem CID 81671
SMILES [Cl].NCC1CC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 205.0°C to 209.0°C
Color White
Elemental Analysis 42.87 to 46.44% C, 12.50 to 13.54% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula C3H5CH2NH2·HCl
Quantity 10 g
Beilstein 12,4
IUPAC Name cyclopropylmethanamine;hydrochloride
Formula Weight 107.58
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (Aminomethyl)cyclopropane hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • (Aminomethyl)cyclopropane hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'