missing translation for 'onlineSavingsMsg'
Learn More
(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals Product Code.: 10732681

(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals

Product Code. 10732681
1 g, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10732681

Brand: Thermo Scientific Acros 291640010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-51-9
Molecular Formula CH6NO3P
Molecular Weight (g/mol) 111.04
MDL Number MFCD00008105
InChI Key MGRVRXRGTBOSHW-UHFFFAOYSA-N
Synonym aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethylShow MoreShow Less
PubChem CID 14017
ChEBI CHEBI:28812
IUPAC Name aminomethylphosphonic acid
SMILES C(N)P(=O)(O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NCH2P(O)(OH)2
Quantity 1 g
Beilstein 01, 2596
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohols
Formula Weight 111.04
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material (Aminomethyl)phosphonic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0394184 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number
Lot NumberA0444776 Certificate TypeCertificate of Analysis Date06/08/2022 Catalog Number
Lot NumberA0409154 Certificate TypeCertificate of Analysis Date03/12/2021 Catalog Number
compliance-icons
Product Identifier
  • (Aminomethyl)phosphonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.