missing translation for 'onlineSavingsMsg'
Learn More

Aminopterin, Thermo Scientific™

Product Code. 17555545 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 17555545

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Packaging Glass Bottle
CAS 54-62-6
Certificate of Analysis Available
Physical Form Powder
Molecular Formula C19H20N8O5
MDL Number 00036692
Color Light Yellow to Yellow/Brown
Melting Point 225°C
Merck Index 13-471
Solubility Soluble in DMF, DMSO
InChI Key TVZGACDUOSZQKY-UHFFFAOYNA-N
SMILES NC1=NC(N)=C2N=C(CNC3=CC=C(C=C3)C(=O)NC(CCC(O)=O)C(O)=O)C=NC2=N1
IUPAC Name 2-[(4-{[(2,4-diaminopteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
Molecular Weight (g/mol) 440.42
Quantity 100 mg
Chemical Name or Material Aminopterin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.