missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, 0.5M solution in 1,4-dioxane, AcroSeal™, Thermo Scientific Chemicals

108.54 € - 589.94 €

Specifications

Chemical Name or Material Ammonia
Color Colorless
Name Note 0.5M solution in 1, 4-dioxane
CAS 7664-41-7, 123-91-1
Molecular Formula H3N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10501892
View Documents
Thermo Scientific Acros
368381000
100 mL AcroSeal™ Glass bottle
108.54 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10127253
View Documents
Thermo Scientific Acros
368380010
1 L AcroSeal™ Glass Bottle
589.94 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonia
0.5M solution in 1, 4-dioxane
H3N
MFCD00011418
05,15
15, 488
Solubility in water: soluble.
N
222
17.03
11°C
Colorless
7664-41-7, 123-91-1
NH3
0.4 to 0.6M
1.0230g/mL
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
QGZKDVFQNNGYKY-UHFFFAOYSA-N
17.03
CHEBI:16134
1.023
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor. Causes serious eye irritation. May cause respiratory irritation. Suspected of causing cancer. Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. Wash face, hands and any exposed skin thoroughly after handling. Use personal protective equipment as required. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

Product Identifier
  • Ammonia, 0.5M solution in 1,4-dioxane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : 1,4-Dioxane, Ammonia

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only