Learn More
Ammonia, 2M in methanol, Thermo Scientific Chemicals
Description
Ammonia, 2M in methanol is used as a solvent in organic reactions and as a mobile phase in thin layer chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Linear Formula | NH3 |
UN Number | UN2924 |
Concentration | 2M in methanol |
Density | 0.787 |
Merck Index | 14,492 |
Solubility Information | Miscible with water. |
Formula Weight | 17.03 |
Quantity | 100 mL |
Flash Point | 6°C (43°F) |
Sensitivity | Hygroscopic |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Safety and Handling




- Ammonia, 2M in methanol
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : methanol, ammonia
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.