missing translation for 'onlineSavingsMsg'
Learn More
Ammonia, 7M in methanol, Thermo Scientific Chemicals Product Code.: 15423917

Ammonia, 7M in methanol, Thermo Scientific Chemicals

Product Code. 15423917
1000 mL
Click to view available options
Quantity:
100 mL
1000 mL
This item is not returnable. View return policy

Product Code. 15423917

Brand: Thermo Scientific Alfa Aesar H30382.AU

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Microwave-assisted synthesis of HX salts of primary amines from their corresponding halides and 7 M ammonia in methanol provides practical high yield access to even volatile primary amines for parallel synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-41-7
Molecular Formula H3N
Molecular Weight (g/mol) 17.03
MDL Number MFCD00011418
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-folShow MoreShow Less
PubChem CID 222
ChEBI CHEBI:16134
IUPAC Name azane
SMILES N

Specifications

Linear Formula NH3
UN Number UN3286
Concentration 7M in methanol
Density 0.779
Merck Index 14,492
Solubility Information Fully misicible in water.
Formula Weight 17.03
Quantity 1000 mL
Flash Point 14°C (57°F)
Sensitivity Hygroscopic
Chemical Name or Material Ammonia
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2226269 Certificate TypeCertificate of Analysis Date07/10/2024 Catalog NumberH30382.AE,H30382.AU
Lot Number2218562 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog NumberH30382.AE,H30382.AU
Lot Number2217013 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog NumberH30382.AE,H30382.AU
Lot Number10249529 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog NumberH30382.AE,H30382.AU
Lot Number10235240 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberH30382.AE,H30382.AU
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonia, 7M in methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : methanol, ammonia

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonia, 7M in methanol, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.