Learn More
WTW™ Ammonia and Ammonium Test Kits
For waste water, drinking water
Brand:  WTW™ 250329
140.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11784745
  • 147.00 € / Pack of 25
Explore more special offers
Description
Description
  • For waste water, drinking water
  • Chromate (chromium VI and total chromium) cell test kit
  • Photometric test method
  • This test is highly accurate and easy to use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Indophenol blue (Berthelot reaction)
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
25
Photometric
For waste water, drinking water, sea water
Certificates
25 Tests
CombiCheck Ammonia (NH3) cell test kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
WTW™ Ammonia and Ammonium Test Kits > Cell Test; Detection Range: 0.5 to 16mg/L NH4-N, 0.6 - 20.6mg/L NH4+